- Formation of γ-oxoacids and 1H-pyrrol-2(5H)-ones from α,β-unsaturated ketones and ethyl nitroacetate.
Formation of γ-oxoacids and 1H-pyrrol-2(5H)-ones from α,β-unsaturated ketones and ethyl nitroacetate.
The Journal of organic chemistry (2010-10-05)
Maialen Aginagalde, Tamara Bello, Carme Masdeu, Yosu Vara, Ana Arrieta, Fernando P Cossío
PMID20886821
ABSTRACT
Michael addition of ethyl nitroacetate on α,β-unsaturated ketones followed by Nef oxidation under hydrolytic conditions yields γ-oxoacids instead of the corresponding α,δ-dioxoesters. A concerted decarboxylation step is proposed on the basis of computational results. Finally, conversion of the γ-ketoacids thus prepared into 1H-pyrrol-2(5H)-ones by reaction with primary amines under Paal-Knorr conditions is also reported.