Merck
  • Cross-linked hyaluronic acid hydrogel films: new biomaterials for drug delivery.
Cross-linked hyaluronic acid hydrogel films: new biomaterials for drug delivery.
Journal of controlled release : official journal of the Controlled Release Society (2000-10-06)
Y Luo, K R Kirker, G D Prestwich
ABSTRACT
A new hyaluronic acid (HA)-based hydrogel film was prepared and evaluated for use in drug delivery. This biocompatible material crosslinks and gels in minutes, and the dried film swells and rehydrates to a flexible hydrogel in seconds. HA was first converted to the adipic dihydrazide derivative and then crosslinked with the macromolecular homobifunctional reagent poly(ethylene glycol)-propiondialdehyde to give a polymer network. After gelation, a solvent casting method was used to obtain a HA hydrogel film. The dried film swelled sevenfold in volume in buffer, reaching equilibrium in less than 100 s. Scanning electron microscopy (SEM) of the hydrogel films showed a condensed and featureless structure before swelling, but a porous microstructure when hydrated. The thermal behavior of the hydrogel films was characterized by differential scanning calorimetry. The enzymatic degradation of the HA hydrogel films by hyaluronidase was studied using both SEM and a spectrophotometric assay. Drug release from the hydrogel film was evaluated in vitro using selected anti-bacterial and anti-inflammatory drugs. This novel biomaterial can be employed for controlled release of therapeutic agents at wound sites.