Merck
Lipase catalyzed synthesis of silicone polyesters.
Chemical communications (Cambridge, England) (2009-11-04)
Yadagiri Poojari, Stephen J Clarson
ABSTRACT
Immobilized Candida antarctica lipase B (CALB) was successfully employed as a catalyst to synthesize silicone aromatic polyesters by the transesterification of dimethyl terephthalate with alpha,omega-bis(hydroxyalkyl)-terminated poly(dimethylsiloxane) in toluene under mild reaction conditions.