Merck
  • Identification of novel transcripts complementary to the Marek's disease virus homologue of the ICP4 gene of herpes simplex virus.
Identification of novel transcripts complementary to the Marek's disease virus homologue of the ICP4 gene of herpes simplex virus.
The Journal of general virology (1994-07-01)
D S Li, J Pastorek, V Zelník, G D Smith, L J Ross
ABSTRACT
Libraries of cDNA were generated from polyadenylated RNAs derived from Marek's disease virus (MDV)-transformed cell lines by directional cloning of oligo-(dT)-primed cDNAs in lambda gt22A. Analysis of the libraries for viral sequences showed that a number of cDNA clones originated from transcripts mapping in the BamHI A region of the MDV genome. Sequencing and fine mapping of these cDNAs suggested that the RNA transcripts expressed from this region were either in the sense or antisense direction with respect to the MDV homologue of the ICP4 gene of herpes simplex virus. The longest cDNA clone from antisense transcripts was 2756 bp long and partially overlapped the 5' end of the coding region of the ICP4 gene. The cDNA clone contained at least four introns, shown by comparison of its sequence with the sequence of the ICP4 gene. The presence of introns was confirmed by PCR analysis. All the introns have the consensus splice donor and acceptor signals at their 5' and 3' ends respectively. Northern blot analysis showed that the ICP4 gene homologue of MDV was abundantly transcribed only in lytically infected fibroblasts, whereas transcripts complementary to the ICP4 gene were the major transcripts in MDV-transformed cell lines and lymphomas. The transcripts complementary to ICP4 consist of two major RNA species approximately 15 kb and 1.32 kb long. The results suggest that there might be an inverse relationship between the abundance of ICP4 transcripts and their complementary transcripts in MDV-infected and transformed cells.