Merck
Modification of phosphatidylserine by hypochlorous acid.
Chemistry and physics of lipids (2009-07-07)
Joerg Flemmig, Holger Spalteholz, Katja Schubert, Sandra Meier, Juergen Arnhold
ABSTRACT
The binding of the heme enzyme myeloperoxidase to phosphatidylserine epitopes on the surface of non-vital polymorphonuclear leukocytes and other cells at inflammatory sites favours modifications of this phospholipid by myeloperoxidase products. As detected by MALDI-TOF mass spectrometry hypochlorous acid and the myeloperoxidase-hydrogen peroxide-chloride system convert 1,2-dipalmitoyl-sn-glycero-3-phosphoserine into 1,2-dipalmitoyl-sn-glycero-3-phosphoacetaldehyde and 1,2-dipalmitoyl-sn-glycero-3-phosphonitrile. A transient chlorimine derivative was detected using 4-chloro-alpha-cyanocinnamic acid as matrix in mass spectrometry only at short incubation times and supplying HOCl in two-fold excess. The decay of transient chlorinated products was followed by changes in absorbance spectra using O-phospho-l-serine to model the behavior of the serine head group in phosphatidylserine. N-Chlorimine and N-monochloramine derivatives decayed with half-life times of 1.5 and 57 min, respectively, at 22 degrees C and pH 7.4. N-Dichloramines decayed within few seconds under these conditions.
Product Description
Supelco
4-Chloro-α-cyanocinnamic acid, ≥95.0% (HPLC)