Merck
  • SPE HG-AAS method for the determination of inorganic arsenic in rice--results from method validation studies and a survey on rice products.
SPE HG-AAS method for the determination of inorganic arsenic in rice--results from method validation studies and a survey on rice products.
Analytical and bioanalytical chemistry (2013-04-23)
Rie R Rasmussen, Yiting Qian, Jens J Sloth
ABSTRACT
The present paper describes the development, validation and application of a method for inorganic arsenic (iAs) determination in rice samples. The separation of iAs from organoarsenic compounds was done by off-line solid-phase extraction (SPE) followed by hydride generation atomic absorption spectrometry (HG-AAS) detection. This approach was earlier developed for seafood samples (Rasmussen et al., Anal Bioanal Chem 403:2825-2834, 2012) and has in the present work been tailored for rice products and further optimised for a higher sample throughput and a lower detection limit. Water bath heating (90 °C, 60 min) of samples with dilute HNO3 and H2O2 solubilised and oxidised all iAs to arsenate (As(V)). Loading of buffered sample extracts (pH 6 ± 1) followed by selective elution of arsenate from a strong anion exchange SPE cartridge enabled the selective iAs quantification by HG-AAS, measuring total arsenic (As) in the SPE eluate. The in-house validation gave mean recoveries of 101-106% for spiked rice samples and in two reference samples. The limit of detection was 0.02 mg kg(-1), and repeatability and intra-laboratory reproducibility were less than 6 and 9%, respectively. The SPE HG-AAS method produced similar results compared to parallel high-performance liquid chromatography coupled to inductively coupled plasma mass spectrometry (ICP-MS) analysis. The SPE separation step was tested collaboratively, where the laboratories (N = 10) used either HG-AAS or ICP-MS for iAs determination in a wholemeal rice powder. The trial gave satisfactory results (HorRat value of 1.6) and did not reveal significant difference (t test, p > 0.05) between HG-AAS and ICP-MS quantification. The iAs concentration in 36 rice samples purchased on the Danish retail market varied (0.03-0.60 mg kg(-1)), with the highest concentration found in a red rice sample.