Merck
  • Adenosine 5-diphosphate antagonists and human platelets: no evidence that aggregation and inhibition of stimulated adenylate cyclase are mediated by different receptors.
Adenosine 5-diphosphate antagonists and human platelets: no evidence that aggregation and inhibition of stimulated adenylate cyclase are mediated by different receptors.
British journal of pharmacology (1982-05-01)
N J Cusack, S M Hourani
ABSTRACT
1 Adenosine 5'-diphosphate (ADP) induces human platelet aggregation and noncompetitively inhibits stimulated human platelet adenylate cyclase; it has been suggested that these two effects are mediated by separate ADP receptors on the platelet surface. 2 Adenosine 5'-triphosphate and seven adenine nucleotide analogues were tested as inhibitors of both effects of ADP on human platelets, and were found to be competitive. 3 pA2 values were calculated for each antagonist for inhibition of both effects of ADP, and a good correlation (correlation coefficient 0.87; p less than 0.01) was found between the pA2 values for inhibition of ADP-induced aggregation and the pA2 values for inhibition of the effect of ADP on stimulated adenylate cyclase. 4 Such a correlation does not support the suggestion that ADP-induced aggregation and the inhibition by ADP of stimulated adenylate cyclase are mediated by two separate receptors.