- Folate antagonists. 12. Antimalarial and antibacterial effects of 2,4-diamino-6-[(aralkyl and alicyclid)thio-, sulfinyl-, and sulfonyl]quinazolines.
Folate antagonists. 12. Antimalarial and antibacterial effects of 2,4-diamino-6-[(aralkyl and alicyclid)thio-, sulfinyl-, and sulfonyl]quinazolines.
Journal of medicinal chemistry (1978-07-01)
E F Elslager, J Davoll, P Jacob, A M Johnson, J Johnson, L M Werbel
PMID97382
ABSTRACT
A series of 2,4-diamino-6-[(aralkyl and alicyclic)thio-, sulfinyl-, and sulfonyl]quinazolines was prepared via condensation of 5-chloro-2-nitrobenzonitrile or 5,6-dichloro-2-nitrobenzonitrile with the appropriate aralkyl or alicyclic thiopseudourea, reduction of the resulting 2-nitro-5-[(aralkyl or alicyclic)thio]benzonitrile with stannous chloride to the amine, and cyclization with chloroformamidine hydrochloride. Oxidation was effected with hydrogen peroxide or the bromine complex of 1,4-diazabicyclo[2.2.2]octane. These analogues when examined for suppressive activity against drug-sensitive lines of Plasmodium berghei in mice were not as active as 2,4-diamino-6-[3,4-dichlorobenzyl)amino]quinazoline (Ia).