Merck
  • Comparison of 16-androstene steroid concentrations in sterile apocrine sweat and axillary secretions: interconversions of 16-androstenes by the axillary microflora--a mechanism for axillary odour production in man?
Comparison of 16-androstene steroid concentrations in sterile apocrine sweat and axillary secretions: interconversions of 16-androstenes by the axillary microflora--a mechanism for axillary odour production in man?
The Journal of steroid biochemistry and molecular biology (1994-03-01)
D B Gower, K T Holland, A I Mallet, P J Rennie, W J Watkins
ABSTRACT
The concentrations of five 16-androstene steroids were determined, by a GC-MS method, in freshly-produced apocrine sweat (adrenaline-induced), in 8 men and 2 women. The ranges of concentrations (nmol/microliter) in apocrine sweat were: 5 alpha-androst-16-en-3-one (5 alpha-A), 0.1-2.0 and 4,16-androstadien-3-one (androstadienone), 0-1.9, 5,16-Androstadien-3 beta-ol (androstadienol) was also found in 5 of the subjects (range 0.05-1.05). 5 alpha-Androst-16-en-3 alpha- or 3 beta-ols [3 alpha (beta)-androstenols] were only found in small amounts (< 0.1 nmol/microliters) in a few subjects. In the second study, prior to apocrine sweat collection (adrenaline injection), the axillary skin of 6 of the male subjects was washed with diethyl ether on an adjacent site of the axillary vault. The concentrations of 16-androstenes were compared in the ethereal extracts and apocrine sweat. The former contained detectable levels (pmol/cm2) of androstadienone (17.9 +/- 2.4), 3 alpha-androstenol (6.9 +/- 3.7), 3 beta-androstenol (1.8 +/- 1.0) and androstadienol (1.9 +/- 0.5) (means +/- SEM) in all 6 subjects. All but 1 subject also had 5 alpha-androstenone, the mean value for the others being 2.5 +/- 0.6. The axillary skin levels of 3 alpha- and 3 beta-androstenols, androstadienol and, in 3 subjects, androstadienone exceeded those in the apocrine sweat obtained from the same subjects, whereas levels of 5 alpha-androstenone in the skin extracts were all lower than in apocrine sweat samples, when related to the corresponding areas of skin sampled. The metabolism of 16-androstenes was studied in vitro in the presence of two aerobic coryneform bacteria, previously shown to metabolize testosterone as well as being capable of producing odour from extracts of axillary sweat in an odour-generation test. Although both coryneforms caused complex metabolic reactions and were capable of oxidation or reduction at C-3 and C-4, the overall direction favoured reduction. For example, large quantities of the more odorous 5 alpha-androstenone and 3 alpha-androstenol were formed from androstadienol and androstadienone. In contrast, strains of corynebacteria, unable to produce odour and incapable of metabolizing testosterone, were also unable to metabolize 16-androstenes.(ABSTRACT TRUNCATED AT 400 WORDS)