Merck
  • Homogentisic acid and structurally related compounds as intermediates in plasma soluble melanin formation and in tissue toxicities.
Homogentisic acid and structurally related compounds as intermediates in plasma soluble melanin formation and in tissue toxicities.
Archives internationales de physiologie, de biochimie et de biophysique (1994-05-01)
Z L Hegedus, U Nayak
ABSTRACT
Homogentisic acid (HGA) spontaneously starts to undergo oxidation and polymerization soon after the beginning of incubation in human blood or plasma at 37 degrees C, and forms plasma soluble melanins (PSM). Haemolysis accompanies this process in blood. The addition of equimolar quantities of antioxidants delays this oxidation significantly (isoascorbic acid by 2:30-4:00 h; glutathione by 3:20-4:05 h; D-penicillamine by 5:00-5:45 h). HGA is a phenylalanine and tyrosine metabolite, related structurally to the catecholamines and other precursors of melanins. HGA is normally metabolized by the enzyme homogentisic acid oxidase. When this enzyme is genetically missing, part of HGA is excreted in the urine, another part polymerizes darkens many tissues (ochronosis), and produces widespread degenerative changes in cartilage and other connective tissues, joints, blood vessels, heart valves, kidneys and in other tissues. Collectively this disorder is known as alcaptonuria, for which no satisfactory treatment is known. The causes of both alcaptonuric arthritis and rheumatoid arthritis are thought to involve increased oxidative stress. Inflammation of joints and connective tissue damage are involved in both diseases. Oxygen radicals are suspected to cause inflammation and cellular damage. Hydroxyl radicals degrade hyaluronic acid (the viscous synovial fluid of joints). High levels of products of free radical reactions, with fluorescence excitation (ex) and emission (em) maxima in the wavelength ranges of those of PSM (ex 320-400 and em 400-470) were reported in the blood sera and synovial fluids of patients with rheumatoid arthritis.(ABSTRACT TRUNCATED AT 250 WORDS)