Merck
  • Urinary 6-keto-prostaglandin F1a in patients with gynaecological tumours.
Urinary 6-keto-prostaglandin F1a in patients with gynaecological tumours.
Cancer letters (1987-02-01)
A Aitokallio-Tallberg, L Viinikka, O Ylikorkala
ABSTRACT
The production of the antiaggregatory and vasodilatory prostacyclin (PGI2) in patients with gynaecological tumours was studied by assaying urinary 6-keto-prostaglandin F1a (= 6-keto-PGF1a), a hydration product of PGI2), by radioimmunoassay following high performance liquid chromatography (HPLC) in 59 patients with gynaecological tumours and 12 non-tumourous control women. Urinary 6-keto-PGF1a excretion in patients with cervical cancer (28.3 +/- 3.6 pmol/mmol creatinine, mean +/- S.E., n = 12), endometrial cancer (22.8 +/- 3.7 pmol/mmol creatinine, n = 12, uterine fibroids (26.0 +/- 3.5 pmol/mmol creatinine, n = 12) benign ovarian cysts (22.4 +/- 1.8 pmol/mmol creatinine, n = 12) did not differ from that in the control women (29.9 +/- 3.6 pmol/mmol creatinine, n = 12). However, patients with ovarian cancer excreted increased amounts of 6-keto-PGF1a (55.4 +/- 10.4 pmol/mmol creatinine, n = 11, P less than 0.05), although this bore no relation to tumour histology, clinical stage or the outcome of the patients. Thus, ovarian cancer is accompanied by increased PGI2 production, perhaps in the kidneys and/or in the cancer tissue.