Merck
  • Quantitative determination of succinylacetone in dried blood spots for newborn screening of tyrosinemia type I.
Quantitative determination of succinylacetone in dried blood spots for newborn screening of tyrosinemia type I.
Molecular genetics and metabolism (2006-02-02)
Mark J Magera, Nishantha D Gunawardena, Si Houn Hahn, Silvia Tortorelli, Grant A Mitchell, Stephen I Goodman, Piero Rinaldo, Dietrich Matern
ABSTRACT
Tyrosinemia type I (TYR 1) is a severe disorder causing early death if left untreated. While tyrosine can be determined in dried blood spots (DBS), it is not a specific marker for TYR 1 and most often associated with benign transient tyrosinemia of the newborn. Succinylacetone (SUAC) is a specific marker for TYR 1 but not detectable by routine newborn screening. We developed a new assay that determines SUAC in DBS by liquid-chromatography tandem mass spectrometry (LC-MS/MS). Whole blood is eluted from a 3/16-in. DBS by an aqueous solution containing deuterium labeled SUAC as internal standard (IS). SUAC and IS are oximated, then extracted, butylated, and analyzed by LC-MS/MS. Quantitation is from SUAC spiked calibrator DBS over the range 0-200 microM using selected reaction monitoring of transitions m/z 212 to 156 and m/z 214 to 140 for SUAC and IS, respectively. Analysis time is 5 min. To assess the effectiveness of a two-tier screening approach for TYR 1 we applied this assay to our newborn screening program over the last 15 months. The intra-assay precision was determined for three different levels of SUAC (5, 20, and 50 micromol/L) and the CV calculated to be 4.7, 2.6, and 3.1%, respectively (n=5). Inter-assay precision CVs were 12.7, 8.2, and 7.8%, respectively on the same samples. SUAC levels in DBS from 10 confirmed TYR 1 cases not treated with 2-(2-nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione (NTBC) were clearly abnormal (16-150 micromol/L; mean: 61 micromol/L; controls: <5 micromol/L). Over a 15-month period, SUAC was determined in newborn screening samples with elevated tyrosine concentrations when applying different cut off values until it was settled at 150 micromol/L. No case of TYR 1 was detected in 124,780 newborns tested. We have developed a new LC-MS/MS based method for the determination of SUAC in DBS. This assay has the potential to significantly reduce the number of false positive results in newborn screening for TYR 1 and can also be used for the laboratory follow up of patients treated for TYR 1.