Merck
  • Retinol, at concentrations greater than the physiological limit, induces oxidative stress and apoptosis in human dermal fibroblasts.
Retinol, at concentrations greater than the physiological limit, induces oxidative stress and apoptosis in human dermal fibroblasts.
Experimental dermatology (2004-03-11)
Amparo Gimeno, Rosa Zaragozá, Inma Vivó-Sesé, Juan R Viña, Vicente J Miralles
ABSTRACT
We have investigated the dose (in the range of microM) and time-dependent effects of four different retinoids (retinol, retinal, retinoic acid and retinol palmitate) on human dermal fibroblasts cultivated in vitro. Retinol and retinal, at a concentration of 20 microM, caused cell damage as evaluated by lactate dehydrogenase activity released into the culture medium. The oxidised glutathione (GSSG)/reduced glutathione (GSH) ratio and malondialdehyde production indicated that 20 microM of retinol provoked oxidative stress in the cultivated human fibroblasts. In the first 8 h after retinol treatment the levels of p53 and Bax proteins as well as caspase 3 activity increased, suggesting apoptotic cell death during the first hours of treatment. If the retinol treatment exceeded 18-24 h we observed necrotic cell death. Vitamin E and coenzyme Q(10) had a protective effect against oxidative stress generated by retinol. Both antioxidant molecules reduced retinol uptake, and in the case of vitamin E the expression of CRABP-II mRNA was induced, providing a plausible explanation for its protective effect.
Product Description
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Retinyl palmitate, potency: ≥1,700,000 USP units per g
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil