- Inhibition of lysophosphatidic acid-induced neurite retraction and cell rounding by SR 57746A.
Inhibition of lysophosphatidic acid-induced neurite retraction and cell rounding by SR 57746A.
Brain research. Molecular brain research (1998-04-29)
M Magazin, P Schiltz, J L Zachayus, E Cavrois, D Caput, P Ferrara
PMID9473704
ABSTRACT
Rapid neurite retraction and transient rounding of serum-starved NG108-15 and PC12 cells by lysophosphatidic acid (LPA) is retarded and reduced by pre-incubation of the cells with the small non-peptidic molecule, SR 57746A, which exhibits neurotrophic properties. The compound also antagonizes the redistribution of filamentous actin by LPA in both cell types. We hypothesize that the SR 57746A attenuation of LPA-induced effects may account for at least some of the neuroprotective properties of this molecule.