Merck
  • Binding and effects of KATP channel openers in the vascular smooth muscle cell line, A10.
Binding and effects of KATP channel openers in the vascular smooth muscle cell line, A10.
British journal of pharmacology (1997-12-24)
U Russ, F Metzger, E Kickenweiz, A Hambrock, P Krippeit-Drews, U Quast
ABSTRACT
1. The ATP-sensitive K+ channel (KATP channel) in A10 cells, a cell line derived from rat thoracic aorta, was characterized by binding studies with the tritiated KATP channel opener, [3H]-P1075, and by electrophysiological techniques. 2. Saturation binding experiments gave a KD value of 9.2 +/- 5.2 nM and a binding capacity (BMax) of 140 +/- 40 fmol mg-1 protein for [3H]-P1075 binding to A10 cells; from the BMax value a density of binding sites of 5-10 per microns2 plasmalemma was estimated. 3. KATP channel modulators such as the openers P1075, pinacidil, levcromakalim and minoxidil sulphate and the blocker glibenclamide inhibited [3H]-P1075 binding. The extent of inhibition at saturation depended on the compound, levcromakalim inhibiting specific [3H]-P1075 binding by 85%, minoxidil sulphate and glibenclamide by 70%. The inhibition constants were similar to those determined in strips of rat aorta. 4. Resting membrane potential, recorded with microelectrodes, was -51 +/- 1 mV. P1075 and levcromakalim produced a concentration-dependent hyperpolarization by up to -25 mV with EC50 values of 170 +/- 40 nM and 870 +/- 190 nM, respectively. The hyperpolarization induced by levcromakalim (3 microM) was completely reversed by glibenclamide with an IC50 value of 86 +/- 17 nM. 5. Voltage clamp experiments were performed in the whole cell configuration under a physiological K+ gradient. Levcromakalim (10 microM) induced a current which reversed around -80 mV; the current-voltage relationship showed considerable outward rectification. Glibenclamide (3 microM) abolished the effect of levcromakalim. 6. Analysis of the noise of the levcromakalim (10 microM)-induced current at -40 and -20 mV yielded estimates of the channel density, the single channel conductance and the probability of the channel to be open of 0.14 micron-2, 8.8 pS and 0.39, respectively. 7. The experiments showed that A10 cells are endowed with functional KATP channels which resemble those in vascular tissue; hence, these cells provide an easily accessible source of channels for biochemical and pharmacological studies. The density of binding sites for [3H]-P1075 was estimated to be one order of magnitude higher than the density of functional KATP channels; assuming a plasmalemmal localization of the binding sites this suggests a large receptor reserve for the openers in A10 cells.