Merck
  • The synergistic purgative action of aloe-emodin anthrone and rhein anthrone in mice: synergism in large intestinal propulsion and water secretion.
The synergistic purgative action of aloe-emodin anthrone and rhein anthrone in mice: synergism in large intestinal propulsion and water secretion.
The Journal of pharmacy and pharmacology (1997-01-01)
T Yagi, K Yamauchi, S Kuwano
ABSTRACT
This study aimed to explore the mechanism involved in the synergistic purgative action of aloe-emodin anthrone and rhein anthrone, the active metabolites of sennoside C. Aloe-emodin anthrone and rhein anthrone, and their equimolar mixture, induced excretion of an approximately equal number of faeces by intracaecal administration at a dose of 23.2 mumol kg-1 in mice (= 1.0 standard dose). The number of wet faeces induced by aloe-emodin anthrone was less than those of rhein anthrone and the mixture. At the same dose, rhein anthrone and the mixture significantly stimulated large intestinal propulsion, though aloe-emodin anthrone had little stimulatory effect. Aloe-emodin anthrone and rhein anthrone decreased net water absorption but could not reverse it to the net secretion at 1/2 dose. The mixture significantly decreased net water absorption and reversed it to the net secretion at this dose. These anthrones did not stimulate mucus secretion in the colon at 1/2 dose. We concluded that the synergistic purgative effect of aloe-emodin anthrone and rhein anthrone in mice results from synergistic stimulation of large intestinal transit and large intestinal water secretion.