Merck
  • Determinants of early and late results of combined valve operations and coronary artery bypass grafting.
Determinants of early and late results of combined valve operations and coronary artery bypass grafting.
The Annals of thoracic surgery (1996-02-01)
W J Flameng, P Herijgers, J Szécsi, P T Sergeant, W J Daenen, I Scheys
ABSTRACT
Factors determining the outcome of operative correction of valvular abnormalities combined with coronary artery bypass grafting are still incompletely defined. Determinants of early and late (more than 90 days) deaths and event-free survival were studied for combined valve operations and coronary artery bypass grafting in 741 patients using multivariate analysis. Ninety-day survival probability was 89% (95% confidence interval, 87% to 92%). Preoperative risk factors for early death were age, female sex, renal failure, New York Heart Association class IV or V, and mitral insufficiency. The operative risk factor was the duration of aortic cross-clamping. Five- and 10-year survival probabilities were 74% (95% confidence interval, 71% to 78%) and 43% (95% confidence interval, 36% to 50%), respectively. Preoperative risk factors for late death were age, preoperative renal failure, New York Heart Association class IV or V, vessel disease, and nonsinus rhythm. Five- and 10-year event-free survival probabilities were 57% (95% confidence interval, 53% to 61%) and 23% (95% confidence interval, 17% to 28%), respectively. Preoperative risk factors for non-event-free survival were age, female sex, reduced left ventricular function, mitral regurgitation, and pacemaker rhythm. The demographic factors of age and female sex; the comorbid condition of renal failure; the cardiac conditions of advanced New York Heart Association class, left ventricular function, mitral regurgitation, vessel disease, and cardiac rhythm; and the operative condition of ischemia time are the most important predictors of clinical outcome after combined valve operations and coronary artery bypass grafting.