- Totally synthetic analogues of siastatin B. III. Trifluoroacetamide analogues having inhibitory activity for tumor metastasis.
Totally synthetic analogues of siastatin B. III. Trifluoroacetamide analogues having inhibitory activity for tumor metastasis.
The Journal of antibiotics (1994-01-01)
Y Nishimura, T Kudo, S Kondo, T Takeuchi, T Tsuruoka, H Fukuyasu, S Shibahara
PMID8119851
ABSTRACT
A trifluoroacetamide analogue of siastatin B, (3S,4S,5R,6R)-6-(trifluoroacetamido)-4,5-dihydroxy-3-piperidine carboxylic acid has been chemically synthesized. This compound, as well as the previously synthesized analogue, (3R,4R,5R,6R)-6-(trifluoroacetamido)-3,4,5-trihydroxy-3-piperid inecarboxylic acid, showed marked inhibitory activity against beta-glucuronidase and significant inhibition of experimental pulmonary metastasis of the highly metastatic melanoma B16.