Merck
  • Transforming activity of selected polycyclic aromatic hydrocarbons and their nitro-derivatives in BALB/3T3 A31-1-1 cells.
Transforming activity of selected polycyclic aromatic hydrocarbons and their nitro-derivatives in BALB/3T3 A31-1-1 cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1994-07-01)
C W Sheu, S N Dobras, I Rodriguez, J K Lee, P P Fu
ABSTRACT
The transforming activities of four polycyclic aromatic hydrocarbons and six of their nitro-derivatives were studied using BALB/3T3 clone A31-1-1 cells in the absence of exogenous metabolic activation. Each compound was assayed two to four times to its maximal level of solubility. A transformation response was induced by 1-nitropyrene, 2-nitropyrene, 4-nitropyrene and benzo[a]pyrene in the BALB/3T3 mouse embryo cells. Pyrene and 7-nitrobenz[a]anthracene produced questionable responses, and benz[a]anthracene, chrysene, 6-nitrobenzo[a]pyrene and 6-nitrochrysene produced negative responses. The capacity of the assay system to indicate tumorigenicity of the test compounds is discussed.