Merck
  • Identification of PDE isozymes in human pulmonary artery and effect of selective PDE inhibitors.
Identification of PDE isozymes in human pulmonary artery and effect of selective PDE inhibitors.
The American journal of physiology (1994-05-01)
K F Rabe, H Tenor, G Dent, C Schudt, M Nakashima, H Magnussen
ABSTRACT
The effects of the nonselective phosphodiesterase (PDE) inhibitor 3-isobutyl-1-methylxanthine (IBMX) and the selective PDE inhibitors motapizone (type III), rolipram (type IV), zardaverine (type III/IV), and zaprinast (type V and I) on prostaglandin F2 alpha (PFG2 alpha)-induced tone in human pulmonary arteries was investigated. Relaxation was achieved by IBMX [concentration eliciting 50% of maximum response (EC50): 11.3 microM, n = 10], motapizone (EC50:3.0 microM, n = 7), zardaverine (EC50: 3.2 microM, n = 9), and zaprinast (EC50: 31.8 microM, n = 6), whereas rolipram was almost ineffective. The combination of motapizone and zaprinast (10 microM) was the most effective relaxant with supra-additive relaxation and a motapizone EC50 of 575 nM. Biochemical studies revealed the presence of the PDE isozymes I, III, IV and V in the cytosolic and particulate phases of arterial homogenates; PDE II was not detectable. Partial inhibition of adenosine 3',5'-cyclic monophosphate (cAMP)-hydrolyzing PDE activity was achieved with rolipram (26 +/- 2.2%) or motapizone (60 +/- 5.4%), whereas there was almost complete inhibition of total PDE activity with zardaverine (81 +/- 2.0%) or the combination of motapizone and rolipram (82 +/- 2.3%). Inhibition of guanosine 3',5'-cyclic monophosphate (cGMP)-hydrolyzing PDE activity was achieved with zaprinast (62 +/- 2.6%) and motapizone (13 +/- 2.3%), indicating the cGMP-hydrolyzing activity of PDE III. We conclude that four out of the five recognized PDE isozyme families are present in human pulmonary artery. PGF2 alpha-induced tone in this tissue is effectively relaxed through PDE inhibitors with selectivity for type III, III/IV, and type V PDE.