Merck
  • Studies on the ionophorous antibiotics. XXVI. The assignments of the 13C-NMR spectra of lonomycin A and mutalomycin.
Studies on the ionophorous antibiotics. XXVI. The assignments of the 13C-NMR spectra of lonomycin A and mutalomycin.
The Journal of antibiotics (1980-09-01)
H Seto, K Mizoue, N Otake
ABSTRACT
All resonances observed in the 13C-nmr spectra of the polyether antibiotics lonomycin A and mutalomycin have been assigned by the aid of biosynthetic method, selective proton decoupling as well as comparison with structurally related compounds.