Merck
  • Influences of dehydroepiandrosterone acetate on ovarian oocytes in mature cycling rats.
Influences of dehydroepiandrosterone acetate on ovarian oocytes in mature cycling rats.
Experientia (1980-11-15)
H Shinohara, S Okoyama, K Akasofu, E Nishida
ABSTRACT
Degeneration of ovarian oocytes occurred to a remarkable extent in rats with polycystic ovaries induced by dehydroepiandrosterone acetate (DHA-Ac) administration. The ratio of degeneration oocytes, compared with the total oocytes examined, finally exceeded 70%.