Merck
Depot fluphenazine: risk/benefit ratio.
The Journal of clinical psychiatry (1984-05-01)
W M Glazer
ABSTRACT
The risks and benefits associated with depot fluphenazine are reassessed by a review and critique of the literature, with an emphasis on controlled studies comparing depot to oral preparations. Specific gaps in our knowledge are noted and recommendations are made for future research.