- [Functional and morphological status of the gastric mucosa mast cells on the administration of dimedrol and suprastin].
[Functional and morphological status of the gastric mucosa mast cells on the administration of dimedrol and suprastin].
Farmakologiia i toksikologiia (1983-01-01)
V M Uspenskiĭ, V A Zelenyĭ, V B Grinevich, A A Fokina
PMID6130970
ABSTRACT
Histochemical and morphometric analysis of mast cells of the gastric and duodenal mucosa of rats given dyphenhydramine and suprastin for a long period of time (up to 45 days) revealed an increase in their functional activity that manifested in the enhancement of the processes of neoformation and maturation of juvenile mast cells and in the increased break down of mature cellular forms.