Merck
  • The effect of anesthetics on hydrogen bonds. An infrared study at low anesthetic concentrations.
The effect of anesthetics on hydrogen bonds. An infrared study at low anesthetic concentrations.
Biophysical chemistry (1985-10-01)
R Buchet, C Sandorfy
ABSTRACT
It is shown that a striking parallelism exists between the anesthetic potency of general halocarbon anesthetics and their influence on the hydrogen bond association constants in N-H...O=C type hydrogen bonds, important for shaping the ion channels. It is further shown that the effect of potent anesthetics (which contain an acidic hydrogen) on the free/associated ratio in such hydrogen bonds is still significant at clinical anesthetic concentrations. It is argued that the results are in keeping with a pluralistic theory of anesthesia based on both hydrophobic and polar interactions.