Merck
  • Neutrophil-mediated injury to endothelial cells. Enhancement by endotoxin and essential role of neutrophil elastase.
Neutrophil-mediated injury to endothelial cells. Enhancement by endotoxin and essential role of neutrophil elastase.
The Journal of clinical investigation (1986-04-01)
L A Smedly, M G Tonnesen, R A Sandhaus, C Haslett, L A Guthrie, R B Johnston, P M Henson, G S Worthen
ABSTRACT
The neutrophil has been implicated as an important mediator of vascular injury, especially after endotoxemia. This study examines neutrophil-mediated injury to human microvascular endothelial cells in vitro. We found that neutrophils stimulated by formyl-methionyl-leucyl-phenylalanine (FMLP), the complement fragment C5a, or lipopolysaccharide (LPS) (1-1,000 ng/ml) alone produced minimal endothelial injury over a 4-h assay. In contrast, neutrophils incubated with endothelial cells in the presence of low concentrations of LPS (1-10 ng/ml) could then be stimulated by FMLP or C5a to produce marked endothelial injury. Injury was maximal at concentrations of 100 ng/ml LPS and 10(-7) M FMLP. Pretreatment of neutrophils with LPS resulted in a similar degree of injury, suggesting that LPS effects were largely on the neutrophil. Endothelial cell injury produced by LPS-exposed, FMLP-stimulated neutrophils had a time course similar to that induced by the addition of purified human neutrophil elastase, and different from that induced by hydrogen peroxide (H2O2). Further, neutrophil-mediated injury was not inhibited by scavengers of a variety of oxygen radical species, and occurred with neutrophils from a patient with chronic granulomatous disease, which produced no H2O2. In contrast, the specific serine elastase inhibitor methoxy-succinyl-alanyl-alanyl-prolyl-valyl-chloromethyl ketone inhibited 63% of the neutrophil-mediated injury and 64% of the neutrophil elastase-induced injury. However, neutrophil-mediated injury was not inhibited significantly by 50% serum, 50% plasma, or purified alpha 1 proteinase inhibitor. These results suggest that, in this system, chemotactic factor-stimulated human neutrophil injury of microvascular endothelial cells is enhanced by small amounts of LPS and may be mediated in large part by the action of neutrophil elastase.