- [Effect of gutimine derivatives on the metamorphosis of Rana temporaria tadpoles].
[Effect of gutimine derivatives on the metamorphosis of Rana temporaria tadpoles].
Ontogenez (1988-01-01)
N Iu Semigolovskiĭ, V V Vasil'eva
PMID3130597
ABSTRACT
A one-time addition of gutimine (0.5 g/l), amtizol (0.2 g/l), and unithiol (0.25 g/l) to the medium at early stages of tadpole development was shown to inhibit metamorphosis only upon addition of unithiol. Repeated addition of gutimine, amtizol, and unithiol to the medium leads to reliable delay in metamorphosis. The delay is more pronounced at addition of the compounds to the medium at late stages of tadpole development (beginning at 40 stage). This suggests that the studied compounds may have anti-thyroid activity. All drugs under study induce the skin clearing.