Merck
  • High power operation of cladding pumped holmium-doped silica fibre lasers.
High power operation of cladding pumped holmium-doped silica fibre lasers.
Optics express (2013-03-14)
Alexander Hemming, Shayne Bennetts, Nikita Simakov, Alan Davidson, John Haub, Adrian Carter
ABSTRACT
We report the highest power operation of a resonantly cladding-pumped, holmium-doped silica fibre laser. The cladding pumped all-glass fibre utilises a fluorine doped glass layer to provide low loss cladding guidance of the 1.95 µm pump radiation. The operation of both single mode and large-mode area fibre lasers was demonstrated, with up to 140 W of output power achieved. A slope efficiency of 59% versus launched pump power was demonstrated. The free running emission was measured to be 2.12-2.15 µm demonstrating the potential of this architecture to address the long wavelength operation of silica based fibre lasers with high efficiency.