Merck
  • Simultaneous determination of NNK and its seven metabolites in rabbit blood by hydrophilic interaction liquid chromatography-tandem mass spectrometry.
Simultaneous determination of NNK and its seven metabolites in rabbit blood by hydrophilic interaction liquid chromatography-tandem mass spectrometry.
Analytical and bioanalytical chemistry (2013-01-12)
Hailei Lang, Sheng Wang, Qidong Zhang, Beibei Zhao, Lei Wang, Baojun Cao, Juan Wang, Jian Mao, Jianxun Zhang
ABSTRACT
A hydrophilic interaction liquid chromatographic-tandem mass spectrometric (HILIC-MS-MS) method for investigation of the in vivo metabolism of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK), a potent carcinogen, in rabbit blood has been developed and validated. This method achieved excellent repeatability and accuracy. Recovery ranged from 76.9 to 116.3 % and precision (as RSD) between 0.53 and 6.52 %. Linearity was good for all compounds (R(2)>0.9990) and the limit of detection (LOD) ranged from 0.016 to 0.082 ng mL(-1). Pharmacokinetic analysis indicated that NNK was rapidly eliminated in vivo in rabbit blood and 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol (NNAL) was the major metabolite. The hydroxy acid, keto acid, and NNAL-N-oxide were also important metabolites in rabbit blood. It is probable that α-methylene hydroxylation was the major pathway of α-hydroxylation of NNK and NNAL in the rabbit.