- [Comparison of different G-CSF treatment effectiveness in experiments on irradiated mice].
[Comparison of different G-CSF treatment effectiveness in experiments on irradiated mice].
Radiatsionnaia biologiia, radioecologiia (2012-12-12)
L M Rozhdestvenskiĭ, R A Shchegoleva, Iu B Deshevoĭ, N I Lisina, B A Titov
PMID23227714
ABSTRACT
In the experiments on F1 (CBA x C57BL) and BALB mice irradiated by 137Cs gamma-rays, preparations of unglycosilated G-SCF such as Neupogen and their domestic analogs Leucostim and Neupomax were investigated. The tests such as 9-day bone marrow cellularity (BMC) and endogenous CFUs, the neutrophile number restoration, the 30-day survival index have shown that all three preparations have an approximately equal effectiveness relating to acute radiation disease treatment and granulopoiesis stimulation after a 5-10 day consecutive administration following irradiation of mice at lethal and sublethal doses. We have come to the conclusion that Leucostim and Neupomax can be regarded as adequate substitutes for Neupogen.