Merck
  • Novel bis-(-)-nor-meptazinol derivatives act as dual binding site AChE inhibitors with metal-complexing property.
Novel bis-(-)-nor-meptazinol derivatives act as dual binding site AChE inhibitors with metal-complexing property.
Toxicology and applied pharmacology (2012-07-31)
Wei Zheng, Juan Li, Zhuibai Qiu, Zheng Xia, Wei Li, Lining Yu, Hailin Chen, Jianxing Chen, Yan Chen, Zhuqin Hu, Wei Zhou, Biyun Shao, Yongyao Cui, Qiong Xie, Hongzhuan Chen
ABSTRACT
The strategy of dual binding site acetylcholinesterase (AChE) inhibition along with metal chelation may represent a promising direction for multi-targeted interventions in the pathophysiological processes of Alzheimer's disease (AD). In the present study, two derivatives (ZLA and ZLB) of a potent dual binding site AChE inhibitor bis-(-)-nor-meptazinol (bis-MEP) were designed and synthesized by introducing metal chelating pharmacophores into the middle chain of bis-MEP. They could inhibit human AChE activity with IC(50) values of 9.63μM (for ZLA) and 8.64μM (for ZLB), and prevent AChE-induced amyloid-β (Aβ) aggregation with IC(50) values of 49.1μM (for ZLA) and 55.3μM (for ZLB). In parallel, molecular docking analysis showed that they are capable of interacting with both the catalytic and peripheral anionic sites of AChE. Furthermore, they exhibited abilities to complex metal ions such as Cu(II) and Zn(II), and inhibit Aβ aggregation triggered by these metals. Collectively, these results suggest that ZLA and ZLB may act as dual binding site AChEIs with metal-chelating potency, and may be potential leads of value for further study on disease-modifying treatment of AD.