Merck
  • A Box-Behnken design for determining the optimum experimental condition of the fungicide (Vapam) sorption onto soil modified with perlite.
A Box-Behnken design for determining the optimum experimental condition of the fungicide (Vapam) sorption onto soil modified with perlite.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2012-05-09)
Seyed N Azizi, Neda Asemi
ABSTRACT
In the present study, response surface methodology (RSM) based on the Box-Behnken design (BBD) was employed to investigate the effects of the different operating conditions on the removal of the fungicide (Vapam) onto soil modified with perlite using sorption process. The process parameters such as pH of the fungicide solution (2, 5 and 8), temperature (15, 25 and 35°C), shaking time (2, 13 and 24 h) and the percentage of perlite in the modified soil (0, 2 and 4 %) were investigated using a four-factor-three-level Box-Behnken design at an initial fungicide concentration of C(0) = 1.6 mg/L as a fixed input parameter. A second-order quadratic model suggested the optimum conditions to be as follows: fungicide solution pH of 3.57, temperature of 15°C, shaking time of 3.5 h and 4% of perlite in the modified soil which resulted in the improvement of Vapam sorption. Under optimum conditions, the fungicide (Vapam) removal was predicted 12.88 μg/g by BBD. The confirmatory experiments were conducted and the results revealed that the fungicide removal was 13.14 μg/g which indicated that the predicted and the observed values of response (Vapam removal) were in close agreement. Therefore, the soil modified with perlite holds good potential for Vapam sorption. This is the first report on fungicide Vapam sorption onto soil modified with perlite using statistical experimental design employing response surface methodology.