- [Screening and detection of nimetazepam and its chief metabolites].
[Screening and detection of nimetazepam and its chief metabolites].
Beitrage zur gerichtlichen Medizin (1990-01-01)
K Schölermann, H Schütz
PMID2241855
ABSTRACT
The article describes analytical data for the screening and detection of nimetazepam and its major metabolites. The methods comprise thin-layer chromatography, gas chromatography, UV-spectrophotometry, infrared- and mass spectrometry.