Merck
  • Aerial ultra-low-volume application of naled: impact on nontarget imperiled butterfly larvae (Cyclargus thomasi bethunebakeri) and efficacy against adult mosquitoes (Aedes taeniorhynchus).
Aerial ultra-low-volume application of naled: impact on nontarget imperiled butterfly larvae (Cyclargus thomasi bethunebakeri) and efficacy against adult mosquitoes (Aedes taeniorhynchus).
Environmental entomology (2011-12-21)
H Zhong, L J Hribar, J C Daniels, M A Feken, C Brock, M D Trager
ABSTRACT
We assessed the exposure and acute toxicity of naled, applied aerially as an ultra-low-volume spray for mosquito control, on late instar larvae of the Miami blue (Cyclargus thomasi bethunebakeri) (Comstock and Huntington 1943) (Lepidoptera: Lycaenidae), an imperiled South Florida butterfly. We concurrently evaluated the control efficacy against caged adult female salt-marsh mosquitoes (Aedes taeniorhynchus) (Wiedemann 1821) (Diptera: Culicidae). This 3-yr study was conducted in north Key Largo (Monroe County, FL) beginning in 2006. The field trials incorporated 15 sampling stations: nine in the target spray zone, three in the spray drift zone at varying distances from the target zone, and three in the control zone not subjected to naled spray drift. A total of six field spray trials were completed, three at an altitude of 30.5 m (100 feet), and three at 45.7 m (150 feet). For all trials, the ultra-low-volume application of Trumpet EC insecticide (78% naled) at a rate of 54.8 ml/ha (0.75 fl. oz/acre) was effective in killing caged adult mosquitoes in the target zone. Butterfly larvae survival was significantly reduced in the spray zone compared with drift and control zones. Analysis of insecticide residue data revealed that the mortality of the late instar butterfly larvae was a result of exposure to excess residues of naled. Additional research is needed to determine mitigation strategies that can limit exposure of sensitive butterflies to naled while maintaining mosquito control efficacy.