Merck
  • Valence anions of N-acetylproline in the gas phase: computational and anion photoelectron spectroscopic studies.
Valence anions of N-acetylproline in the gas phase: computational and anion photoelectron spectroscopic studies.
The Journal of chemical physics (2011-09-29)
Lidia Chomicz, Janusz Rak, Piotr Paneth, Michael Sevilla, Yeon Jae Ko, Haopeng Wang, Kit H Bowen
ABSTRACT
We report the photoelectron spectrum of anionic N-acetylproline, (N-AcPro)(-), measured with 3.49 eV photons. This spectrum, which consists of a band centered at an electron binding energy of 1.4 eV and a higher energy spectral tail, confirms that N-acetylproline forms a valence anion in the gas phase. The neutrals and anions of N-AcPro were also studied computationally at the B3LYP∕6-31++G(d,p) level. Based on the calculations, we conclude that the photoelectron spectrum is due to anions which originated from proton transfer induced by electron attachment to the π* orbital localized at the acetyl group of N-AcPro. We also characterized the energetics of reaction paths leading to pyrrolidine ring opening in the anionic N-AcPro. These data suggest that electron induced decomposition of peptides/proteins comprising proline strongly depends on the presence of proton donors in the close vicinity to the proline residue.