Merck
  • The influence of the peptide bond on the conformation of amino acids: a theoretical and FT-IR matrix-isolation study of N-acetylproline.
The influence of the peptide bond on the conformation of amino acids: a theoretical and FT-IR matrix-isolation study of N-acetylproline.
Biophysical chemistry (2011-08-16)
Bram Boeckx, Riet Ramaekers, Guido Maes
ABSTRACT
A combined experimental matrix-isolation FT-IR and theoretical study has been performed to investigate the conformational behavior of N-acetylproline. The conformational landscape of N-acetylproline was explored using successively higher computational methods, i.e. HF, DFT(B3LYP) and finally MP2. The exploration resulted in 10 conformations with a relative energy difference smaller than 22 kJ.mol(-1) at the HF/3-21G level of theory. These conformations led to six different conformations after DFT(B3LYP) optimizations. Further optimization at the MP2/6-31++G** level of theory resulted in the same six conformations, all of them with an energy difference smaller than 11.5kJ.mol(-1). One conformation with an intramolecular H-bond was found which was energetically the most favorable conformation. The vibrational and thermodynamical features were calculated using the DFT and MP2 methodologies. In the experimental matrix-isolation FT-IR spectrum, the most stable conformation was dominant and at least two non-H-bonded conformations could be identified. An experimental rotamerization constant between the H-bonded and the other non-H-bonded conformations was estimated and appeared to agree reasonably well with the theoretical MP2 predictions. Some new spectral features of N-acetylproline compared to proline were discovered which might be used to discriminate between the acetylated and non-acetylated form.