Merck
  • Triterpenoid pristimerin synergizes with taxol to induce cervical cancer cell death through reactive oxygen species-mediated mitochondrial dysfunction.
Triterpenoid pristimerin synergizes with taxol to induce cervical cancer cell death through reactive oxygen species-mediated mitochondrial dysfunction.
Anti-cancer drugs (2011-06-07)
Da-Young Eum, Joo-Yun Byun, Chang-Hwan Yoon, Woo-Duck Seo, Ki-Hun Park, Jin-Hwan Lee, Hee Young Chung, Sungkwan An, Yongjoon Suh, Min-Jung Kim, Su-Jae Lee
ABSTRACT
A combined treatment with conventional chemotherapies can enhance the effectiveness of chemotherapeutic agents against cancers. Here, we have shown that the naturally occurring triterpenoids synergistically enhance the response of cervical cancer cells to taxol. Of the triterpenoid compounds, pristimerin enhanced the anticancer effect of taxol with the highest efficiency by combination. Pristimerin synergizes with taxol to inhibit clonogenic survival and tumor growth in nude mice, and to enhance cell death in cervical cancer cells. A combined treatment with taxol and pristimerin induced cervical cancer cell death by increasing intracellular reactive oxygen species levels, upregulation of death receptor death receptor 5 (DR5), activation of Bax, and dissipation of mitochondrial membrane potential. Treatment with N-acetyl-L-cysteine, a thiol-containing antioxidant completely blocked combined treatment-induced Bax translocation as well as DR5 upregulation. Moreover, inhibition of Jun N-terminal kinase/c-Jun pathway attenuated cell death by blocking DR5 upregulation and Bax activation. These results indicate that the triterpenoid, pristimerin, synergistically enhances taxol response of cervical cancer cells through DR5 expression and Bax activation. Furthermore, the reactive oxygen species-dependent activation of the Jun N-terminal kinase/c-Jun pathway is required for the DR5 upregulation and Bax activation. The molecular mechanism revealed by this study may aid in the design of future combination cancer therapies against cells with intrinsically reduced sensitivity to taxol.