Merck
  • Correlation between improvement in functional signs and plethysmographic parameters during venoactive treatment (Cyclo 3 Fort).
Correlation between improvement in functional signs and plethysmographic parameters during venoactive treatment (Cyclo 3 Fort).
International angiology : a journal of the International Union of Angiology (2011-05-28)
F A Allaert, C Hugue, M Cazaubon, J M Renaudin, T Clavel, P Escourrou
ABSTRACT
The aim of this study was to compare the change in functional signs of venous insufficiency and venous refilling time measured by mercury strain-gauge plethysmography under the effects of the combination of Ruscus aculeatus, hesperidin methylchalcone and ascorbic acid (Cyclo 3 Fort). An open-label clinical trial conducted in 65 women presenting with CEAP class C2s and C3s assessment criteria. At D0 and D28, functional signs were measured on a visual analog scale (VAS), venous refilling time by mercury strain-gauge plethysmography and venous reflux by echo-Doppler method. Under treatment, venous refilling time increased from 11.7±4 s to 13.8±4.4 s with Δ of 26% (P<0.0001) and the percentage of patients showing improvement of functional symptoms ≥30% was significantly correlated (P=0.04) with the percentage of patients presenting an improvement in venous refilling time ≥10%. Improvement in subjective functional signs under treatment with Cyclo 3 Fort was correlated with objective plethysmographic parameter improvement. There-fore, this study contributes to objectively document the benefit of prescribing this venoactive drug to active women with CEAP class C2s to C3s.