- Anticoagulation and antiplatelet effects of semax under conditions of acute and chronic immobilization stress.
Anticoagulation and antiplatelet effects of semax under conditions of acute and chronic immobilization stress.
Bulletin of experimental biology and medicine (2010-11-30)
M E Grigorjeva, L A Lyapina
PMID21113455
ABSTRACT
The effects of semax on anticoagulant, fibrinolytic, and platelet components of the anticoagulation system were studied on albino rats under conditions of acute and chronic immobilization stress. Semax exhibited a protective antistress effect after repeated intranasal administration under conditions of hypercoagulation developing in response to immobilization stress of different degree. The effect manifested in stimulation of the anticoagulation system.