Merck
  • A scanning electron microscopic investigation of bonding of methacryloyloxyalkyl hydrogen maleate to etched dentin.
A scanning electron microscopic investigation of bonding of methacryloyloxyalkyl hydrogen maleate to etched dentin.
Journal of dental research (1990-01-01)
T Fukushima, T Horibe
ABSTRACT
Three types of methacryloyloxyalkyl hydrogen maleates--2-(methacryloyloxy)ethyl hydrogen maleate (2MEM), 1-(methacryloyloxy-methyl)ethyl hydrogen maleate (MMEM), and 1-(methacryloyloxymethyl)propyl hydrogen maleate (MMPM)--were prepared for use as bonding agents. The relationships between the thickness of an acid-proof dentin layer and the bond strength and chemical structure of the synthesized monomers were investigated. The bond strengths of 2MEM/composite system to dentin treated with either 37% phosphoric acid solution or 10% citric acid-3% ferric chloride solution (10-3 solution) were superior to those of the others. The acid-proof dentin layer for all bonding agents containing the synthesized monomers at the resin-dentin interface was clearly visible in a scanning electron microscope, regardless of the types of dentin etching agents used. After treatment of the dentin with the phosphoric acid solution, the layers of 2MEM, MMEM, and MMPM were 5-6 micron, 3-4 micron, and 2-3 micron thick, respectively. When the dentin was treated with the 10-3 solution, the layer for each bonding agent was approximately 1 micron thick.