- Etofenamate associated with Lyell syndrome: a case report.
Etofenamate associated with Lyell syndrome: a case report.
Clinical toxicology (Philadelphia, Pa.) (2010-07-01)
Yalcin Golcuk, Deniz Oray, Ozge Duman Atilla, Nilay Tefennioglu
PMID20586575
ABSTRACT
Toxic epidermal necrolysis (Lyell syndrome) is a rare, acute, and potentially life-threatening mucocutaneous disease that is most often triggered by drugs. This is the first case of toxic epidermal necrolysis because of treatment with etofenamate of which we are aware.