Merck
  • Plumbagin activates ERK1/2 and Akt via superoxide, Src and PI3-kinase in 3T3-L1 cells.
Plumbagin activates ERK1/2 and Akt via superoxide, Src and PI3-kinase in 3T3-L1 cells.
European journal of pharmacology (2010-04-28)
Su-Jung Yang, Shi-Chuan Chang, Hui-Chin Wen, Ching-Yu Chen, Jyh-Fei Liao, Chung-Ho Chang
ABSTRACT
Plumbagin, derived from the plant Plumbago zeylanica, has been shown to chronically activate ERK1/2 and inhibit Akt activity in cancer cells. However, the acute effects of plumbagin on ERK1/2 and Akt activities remain unknown. In this study, we examined the effects of plumbagin on ERK1/2 and Akt activities in 3T3-L1 cells. Exposure of 3T3-L1 cells to plumbagin generated superoxide and activated both ERK1/2 and Akt. The plumbagin-stimulated ERK1/2 and Akt activities were sensitive to an antioxidant NAC, superoxide dismutase mimetic MnTBAP, superoxide scavenger Tiron and NAD(P)H oxidase inhibitor DPI. Plumbagin-stimulated ERK1/2 activity was attenuated by the MEK1/2 inhibitor PD98059 and Ras inhibitor manumycin A, whereas plumbagin-stimulated Akt activity was blocked by the PI3K inhibitor LY294002. Both plumbagin-stimulated ERK1/2 and Akt activities were attenuated by PP2, a Src inhibitor. Interestingly, inhibition of phosphatidylinositol 3-kinase (PI3-kinase), but not Akt, activity leaded to attenuation of plumbagin-stimulated ERK1/2 activity. These results suggest that plumbagin activates NAD(P)H oxidase, Src, and PI3K, and that the activated PI3K or PDK1 subsequently stimulate Akt and Ras-Raf-MEK1/2-ERK1/2 in 3T3-L1 cells.