- How to predict product yield in kinetically controlled enzymatic peptide synthesis.
How to predict product yield in kinetically controlled enzymatic peptide synthesis.
Biotechnology and bioengineering (1992-07-01)
M Y Gololobov, E V Kozlova, I L Borisov, U Schellenberger, V Schellenberger, H D Jakubke
PMID18601134
ABSTRACT
The published theory of kinetically controlled enzymatic peptide synthesis needed experimental verification. We carried out the measurement of the peptide yield and estimation of the key parameters alpha and beta for papain-catalyzed synthesis of Mal-L-Phe-L-Ala-LLeuNH(2) from Mal-L-Phe-L-AlaOMe and L-LeuNH(2). The experimental results demonstrate that this theory adequately describes the real process.
MATERIALS