- Botulinum neurotoxin A protease: discovery of natural product exosite inhibitors.
Botulinum neurotoxin A protease: discovery of natural product exosite inhibitors.
Journal of the American Chemical Society (2010-02-18)
Peter Silhár, Katerina Capková, Nicholas T Salzameda, Joseph T Barbieri, Mark S Hixon, Kim D Janda
PMID20158239
ABSTRACT
A new mechanistic class of BoNT/A zinc metalloprotease inhibitors, from Echinacea, exemplified by the natural product d-chicoric acid (I1) is disclosed. A detailed evaluation of chicoric acid's mechanism of inhibition reveals that the inhibitor binds to an exosite, displays noncompetitive partial inhibition, and is synergistic with a competitive active site inhibitor when used in combination. Other components found in Echinacea, I3 and I4, were also inhibitors of the protease.