Merck
  • In vitro and in vivo evaluation of self-microemulsifying drug delivery system of buparvaquone.
In vitro and in vivo evaluation of self-microemulsifying drug delivery system of buparvaquone.
Drug development and industrial pharmacy (2010-02-09)
Gantala Venkatesh, M I A Majid, S M Mansor, N K Nair, Simon L Croft, V Navaratnam
ABSTRACT
The aim of this study was to prepare a lipid-based self-microemulsifying drug delivery system (SMEDDS) to increase the solubility and oral bioavailability of a poorly water-soluble compound, buparvaquone (BPQ). The solubility of BPQ was determined in various vehicles, and pseudo-ternary phase diagrams were constructed to determine the microemulsion region. A series of formulations with different compositions were selected in the microemulsion region for assessment of self-emulsification time and droplet size. The optimized SMEDDS formulation was used for in vitro dissolution and pharmacokinetic studies in rabbits. The optimum formulation of SMEDDS consisted of Capryol 90 (9.82%), Cremophor EL (70.72%), Labrasol (17.68%), and BPQ (1.78%). Emulsification time and the mean droplet size were found to be 1 minute and 18.0 +/- 0.25 nm, respectively, for the optimum formulation. The cumulative percentage of drug released in 90 minutes was 100% in both SGF and SIF. The calculated absolute oral bioavailability for BPQ was found to be 40.10%. The optimum SMEDDS formulation was increased the rate and extent of absorption of BPQ. The formulation is suitable for oral administration of BPQ. It would be useful to conduct efficacy studies of BPQ in diseased animal models and subsequently for toxicokinetics studies.