- Behaviour studies of the fungicide cymoxanil in two strains of the fungus Botrytis cinerea and in haemolymph of locust and lobster. I. In situ monitoring by internal surface reversed-phase high-performance liquid chromatography.
Behaviour studies of the fungicide cymoxanil in two strains of the fungus Botrytis cinerea and in haemolymph of locust and lobster. I. In situ monitoring by internal surface reversed-phase high-performance liquid chromatography.
Journal of chromatography (1991-05-31)
J C Cherton, M T Chevassus-Clement, C Lange, O Convert, R Fritz, P Leroux
PMID1939461
ABSTRACT
A method for following the metabolism of the fungicide cymoxanil in various biological media is described. By using a recently developed high-performance liquid chromatographic method, with an internal surface reversed-phase column, it is unnecessary to clean up the sample before analysis. Thus this technique makes monitoring in fungi as well as in arthropod haemolymph easier and faster.