Merck
  • LPS-induced MCP-1 expression in human microvascular endothelial cells is mediated by the tyrosine kinase, Pyk2 via the p38 MAPK/NF-kappaB-dependent pathway.
LPS-induced MCP-1 expression in human microvascular endothelial cells is mediated by the tyrosine kinase, Pyk2 via the p38 MAPK/NF-kappaB-dependent pathway.
Molecular immunology (2008-10-29)
Appakkudal R Anand, Ritu Bradley, Ramesh K Ganju
ABSTRACT
Bacterial endotoxin (lipopolysaccharide or LPS) has potent pro-inflammatory properties and acts on many cell types including endothelial cells. Secretion of the CC chemokine, MCP-1 (CCL2) by LPS-activated endothelial cells contributes substantially to the pathogenesis of sepsis. However, the mechanism involved in LPS-induced MCP-1 production in endothelial cells is not well understood. Using human microvascular endothelial cells (HMVEC), we analyzed the involvement of the non-receptor tyrosine kinase, Pyk2, in LPS-mediated MCP-1 production. There was a marked activation of the non-receptor tyrosine kinase, Pyk2, in response to LPS. Inhibition of Pyk2 activity using a pharmacological inhibitor, Tyrphostin A9 significantly attenuated LPS-induced Pyk2 tyrosine phosphorylation, p38 MAP kinase (MAPK) activation, NF-kappaB activation, and MCP-1 expression. Furthermore, specific inactivation of Pyk2 activity by transducing microvascular endothelial cells with catalytically inactive Pyk2 mutant (AAV-Pyk2MT) or Pyk2-specific siRNA significantly blocked LPS-induced MCP-1 production. The supernatants of these LPS-stimulated cells with attenuated Pyk2 activity demonstrated decreased trans-endothelial monocyte migration in comparison to LPS-treated controls, thus confirming the inhibition of functional MCP-1 production. In summary, our data suggest a critical role for the Pyk2 mediated pathway involving p38 MAP kinase and NF-kappaB in LPS-induced MCP-1 production in human microvascular endothelial cells.