Merck
Arcaine and MK-801 make recall state-dependent in rats.
Psychopharmacology (2008-09-02)
Ana Paula Chiapinotto Ceretta, Keli Camera, Carlos Fernando Mello, Maribel Antonello Rubin
ABSTRACT
The polyamines putrescine, spermidine, and spermine are a group of aliphatic amines that may act as physiological modulators of the N-methyl-D-aspartate (NMDA) receptor, a glutamate receptor implicated in memory formation and consolidation. Arcaine is a competitive antagonist of the polyamine binding site at the NMDA receptor, the post-training administration of which impairs memory of various tasks. In this study, we investigated whether the administration of arcaine and MK-801 alters the memory of the step-down inhibitory avoidance task, and whether the effects of these NMDA antagonists involve state-dependency mechanisms, in adult male Wistar rats. The administration of arcaine (30 mg/kg, i.p.) or MK-801 (0.03 mg/kg, i.p.) immediately after training impaired inhibitory avoidance performance at testing. Arcaine- and MK-801-induced performance impairment was reversed by the administration of arcaine (30 mg/kg, i.p.) and MK-801 (0.03 mg/kg, i.p.), respectively, 30 min before testing. Response transfer also occurred if arcaine substituted MK-801 at testing, and vice-versa. These results suggest that arcaine and MK-801 induce state-dependent recall and that, probably due to their ability to decrease NMDA receptor function, one drug can substitute for the other at testing, demonstrating a cross-state dependency between arcaine and MK-801.