Merck
  • An enantioselective synthetic route to cis-2,4-disubstituted and 2,4-bridged piperidines.
An enantioselective synthetic route to cis-2,4-disubstituted and 2,4-bridged piperidines.
The Journal of organic chemistry (2008-08-01)
Mercedes Amat, Maria Pérez, Annamaria T Minaglia, Joan Bosch
ABSTRACT
A synthetic route to enantiopure cis-2,4-disubstituted and 2,4-bridged piperidines is reported, the key step being a stereoselective conjugate addition of an organocuprate to a phenylglycinol-derived unsaturated lactam bearing a substituent at the 8a-position.