- Spectroscopic studies of 1,2-diaminoanthraquinone (DAQ) as a fluorescent probe for the imaging of nitric oxide in living cells.
Spectroscopic studies of 1,2-diaminoanthraquinone (DAQ) as a fluorescent probe for the imaging of nitric oxide in living cells.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2008-01-03)
Francisco Galindo, Nurul Kabir, Jelena Gavrilovic, David A Russell
PMID18167606
ABSTRACT
Spectroscopic analysis of the fluorescent probe 1,2-diaminoanthraquinone () provides information about the mechanism of nitric oxide imaging in living cells. Fluorescent aggregates of a reaction product of are thought to be responsible for the images obtained with confocal fluorescence microscopy.